Novus Biologicals
Manufacturer Code:NBP232365
Catalog # NBP232365
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: TGHWNGQQAPPDAGFPVVCCEDVFLSDPLLPRGQRVPLYLSKAPQQMMGSL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 25 kDa Sin3-associated polypeptide; 25 KDa Sin3-Associated Polypeptide Histone Deacetylase Complex Subunit SAP25 Sin3 Corepressor Complex Subunit SAP25 Sin3A-Associated Protein 25kDa Sin3A-Binding Protein 25 Sin3-Associated Polypeptide 25kDa; Histone deacetylase complex subunit SAP25; Sin3 corepressor complex subunit SAP25; sin3-associated polypeptide, 25kDa; Sin3A associated protein 25kDa; Sin3A-associated protein, 25kDa; sin3A-binding protein 25
Gene Aliases: SAP25
UniProt ID: (Human) Q8TEE9
Entrez Gene ID: (Human) 100316904
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.