Invitrogen
Manufacturer Code:MA527707
Catalog # PIMA527707
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S19-2 |
Immunogen | Fusion protein amino acids 1-120 of rat SAP102 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: discs large homolog 3; discs, large homolog 3; Disks large homolog 3; DLG3 MRX90 NEDLG XLMR Disks large homolog 3 PSD-95/SAP90-related protein 1 Synapse-associated protein 102 SAP-102; Neuroendocrine-DLG; protein phosphatase 1, regulatory subunit 82; PSD-95/SAP90-related protein 1; SAP-102; SAP102; Synapse-associated protein 102; XLMR
Gene Aliases: DLG3; Dlgh3; KIAA1232; mKIAA1232; MRX; MRX90; NEDLG; PPP1R82; SAP102; XLMR
UniProt ID: (Human) Q92796, (Rat) Q62936, (Mouse) P70175
Entrez Gene ID: (Human) 1741, (Rat) 58948, (Mouse) 53310
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.