Novus Biologicals
Manufacturer Code:NBP121672B
Catalog # NBP121672B
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S19-2 |
Immunogen | Fusion protein amino acids 1-120 of rat SAP102 |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: discs large homolog 3; discs large homolog 3 (Drosophila) KIAA1232discs large homolog 3 (neuroendocrine-dlg Drosophila) MRX90MRX NEDLGdisks large homolog 3 neuroendocrine-DLG SAP-102 SAP102NE-Dlg Synapse-associated protein 102 XLMR; discs, large homolog 3; Disks large homolog 3; Neuroendocrine-DLG; protein phosphatase 1, regulatory subunit 82; SAP-102; Synapse-associated protein 102; XLMR
Gene Aliases: DLG3; KIAA1232; MRX; MRX90; NEDLG; PPP1R82; SAP102; XLMR
UniProt ID: (Human) Q92796
Entrez Gene ID: (Human) 1741
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.