Novus Biologicals
Manufacturer Code:NBP273985F
Catalog # NBP273985F
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3F2 |
Immunogen | Full length recombinant protein of human SAP102 (NP_066943) produced in HEK293T cells. |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: discs large homolog 3; discs large homolog 3 (Drosophila) KIAA1232discs large homolog 3 (neuroendocrine-dlg Drosophila) MRX90MRX NEDLGdisks large homolog 3 neuroendocrine-DLG SAP-102 SAP102NE-Dlg Synapse-associated protein 102 XLMR; discs, large homolog 3; Disks large homolog 3; Neuroendocrine-DLG; protein phosphatase 1, regulatory subunit 82; SAP-102; Synapse-associated protein 102; XLMR
Gene Aliases: DLG3; KIAA1232; MRX; MRX90; NEDLG; PPP1R82; SAP102; XLMR
UniProt ID: (Human) Q92796
Entrez Gene ID: (Human) 1741
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.