Invitrogen
Manufacturer Code:PA578500
Catalog # PIPA578500
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of Human SAP102. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: discs large homolog 3; Disks large homolog 3; Dlg3 Dlgh3 SAP102 SAP102 discs large homolog 3 (Drosophila) mKIAA1232; PSD-95/SAP90-related protein 1; SAP-102; SAP102; Synapse-associated protein 102
Gene Aliases: Dlg3; Dlgh3; mKIAA1232; SAP102
UniProt ID: (Rat) Q62936, (Mouse) P70175
Entrez Gene ID: (Rat) 58948, (Mouse) 53310
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.