Invitrogen
Manufacturer Code:PA568165
Catalog # PIPA568165
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthesized peptide derived from human SAP102. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: discs large homolog 3; discs, large homolog 3; Disks large homolog 3; MRX MRX90 NEDLG SAP102 XLMR disks large homolog 3 neuroendocrine-DLG synapse-associated protein 102; Neuroendocrine-DLG; protein phosphatase 1, regulatory subunit 82; PSD-95/SAP90-related protein 1; SAP-102; SAP102; Synapse-associated protein 102; XLMR
Gene Aliases: DLG3; Dlgh3; KIAA1232; mKIAA1232; MRX; MRX90; NEDLG; PPP1R82; SAP102; XLMR
UniProt ID: (Human) Q92796, (Rat) Q62936, (Mouse) P70175
Entrez Gene ID: (Human) 1741, (Rat) 58948, (Mouse) 53310
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.