Invitrogen
Manufacturer Code:PA531202
Catalog # PIPA531202
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 290 of Human SAMM50 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: OMP85 SAM50 TOB55 TRG-3 YNL026W sorting and assembly machinery 50kDa sorting and assembly machinery component 50 homolog transformation-related gene 3 protein; sorting and assembly machinery 50kDa; Sorting and assembly machinery component 50 homolog; Transformation-related gene 3 protein; TRG-3
Gene Aliases: 1110030L07Rik; CGI-51; OMP85; SAM50; SAMM50; TOB55; TRG-3; TRG3; YNL026W
UniProt ID: (Human) Q9Y512, (Mouse) Q8BGH2
Entrez Gene ID: (Human) 25813, (Mouse) 68653
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.