Invitrogen
Manufacturer Code:MA525316
Catalog # PIMA525316
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3F5 |
Immunogen | Full length human recombit protein of SAMHD1 produced in HEK293T cell |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: DCIP; Dendritic cell-derived IFNG-induced protein; Deoxynucleoside triphosphate triphosphohydrolase SAMHD1; dNTPase; hSAMHD1; IFN-gamma induced; Interferon-gamma-inducible protein Mg11; Monocyte protein 5; MOP-5; mSAMHD1; RP1-132F21.1 CHBL2 DCIP HDDC1 MOP-5 SBBI88 SAM domain and HD domain-containing protein 1 dendritic cell-derived IFNG-induced protein monocyte protein 5; SAM domain and HD domain 1; SAM domain and HD domain-containing protein 1
Gene Aliases: CHBL2; DCIP; E330031J07Rik; EGK_02438; HDDC1; Mg11; Mg21; MOP-5; MOP5; SAMHD1; SBBI88
UniProt ID: (Human) Q9Y3Z3, (Mouse) Q60710
Entrez Gene ID: (Rhesus monkey) 709060, (Human) 25939, (Dog) 485862, (Rat) 311580, (Mouse) 56045
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.