Novus Biologicals
Manufacturer Code:NBP203285
Catalog # NBP203285
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3F5 |
Immunogen | Full length human recombinant protein of human SAMHD1(NP_056289) produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: DCIP; DCIPMg11 Dendritic cell-derived IFNG-induced protein EC 3.1.4.- HDDC1 Monocyte protein 5 MOP5 MOP-5AGS5 SAM domain and HD domain 1 SAM domain and HD domain-containing protein 1 SBBI88; Dendritic cell-derived IFNG-induced protein; Deoxynucleoside triphosphate triphosphohydrolase SAMHD1; dNTPase; hSAMHD1; Monocyte protein 5; MOP-5; SAM domain and HD domain 1; SAM domain and HD domain-containing protein 1
Gene Aliases: CHBL2; DCIP; HDDC1; MOP-5; MOP5; SAMHD1; SBBI88
UniProt ID: (Human) Q9Y3Z3
Entrez Gene ID: (Human) 25939
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.