Proteintech
Manufacturer Code:125861AP150UL
Catalog # 50-555-004
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SAMHD1 Fusion Protein Ag3287 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DCIP; DCIP HDDC1 Monocyte protein 5 MOP 5 MOP5 SAM domain and HD domain 1 SAMHD1 SBBI88; Dendritic cell-derived IFNG-induced protein; Deoxynucleoside triphosphate triphosphohydrolase SAMHD1; dNTPase; hSAMHD1; IFN-gamma induced; Interferon-gamma-inducible protein Mg11; Monocyte protein 5; MOP-5; mSAMHD1; SAM domain and HD domain 1; SAM domain and HD domain-containing protein 1
Gene Aliases: CHBL2; DCIP; E330031J07Rik; HDDC1; Mg11; Mg21; MOP-5; MOP5; SAMHD1; SBBI88
UniProt ID: (Human) Q9Y3Z3, (Mouse) Q60710
Entrez Gene ID: (Human) 25939, (Rat) 311580, (Mouse) 56045
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.