Invitrogen
Manufacturer Code:PA571622
Catalog # PIPA571622
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 38-67 amino acids from the N-terminal region of human SAMD8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ceramide phosphoethanolamine synthase; CPE synthase; epididymis luminal protein 177; epididymis secretory sperm binding protein Li 181mP; GrailEXP prediction; HEL-S-181mP; SAM domain-containing protein 8; SMSr; sphingomyelin synthase related; Sphingomyelin synthase-related protein 1; Sphingomyelin synthase-related protein 1 SMSr 278- Ceramide phosphoethanolamine synthase CPE synthase Sterile alpha motif domain-containing protein 8 SAM domain-containing protein 8 SAMD8; sterile alpha motif domain-containing 8; Sterile alpha motif domain-containing protein 8
Gene Aliases: 1110053F04Rik; 1700010P07Rik; HEL-177; SAMD8; SMSr
UniProt ID: (Human) Q96LT4, (Mouse) Q9DA37
Entrez Gene ID: (Human) 142891, (Mouse) 67630
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.