Invitrogen
Manufacturer Code:PA5114138
Catalog # PIPA5114138
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human SAMD8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ceramide phosphoethanolamine synthase; Ceramide phosphoethanolamine synthase CPE synthase epididymis luminal protein 177 epididymis secretory sperm binding protein Li 181mP FLJ25082 HEL-177 HEL-S-181mP RP11-487I5.5 SAM domain-containing protein 8 SAMD8 SMSr sphingomyelin synthase related Sphingomyelin synthase-related protein 1 sterile alpha motif domain containing 8 Sterile alpha motif domain-containing protein 8; CPE synthase; epididymis luminal protein 177; epididymis secretory sperm binding protein Li 181mP; HEL-S-181mP; SAM domain-containing protein 8; SMSr; sphingomyelin synthase related; Sphingomyelin synthase-related protein 1; Sterile alpha motif domain-containing protein 8
Gene Aliases: HEL-177; SAMD8; SMSr
UniProt ID: (Human) Q96LT4
Entrez Gene ID: (Human) 142891
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.