Proteintech
Manufacturer Code:173871AP150UL
Catalog # 50-173-1543
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SAMD4A Fusion Protein Ag11273 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hSmaug1 KIAA1053 Protein Smaug homolog 1 SAMD4 SAMD4A Smaug Smaug 1 Smaug1 SMG SMGA; Protein Smaug homolog 1; SAM domain-containing protein 4A; Smaug 1; sterile alpha motif domain containing 4; Sterile alpha motif domain-containing protein 4A
Gene Aliases: 1700024G08Rik; 1700111L17Rik; 4933436G17Rik; KIAA1053; SAMD4; SAMD4A; SMAUG; SMAUG1; SMG; SMGA
UniProt ID: (Human) Q9UPU9, (Mouse) Q8CBY1
Entrez Gene ID: (Human) 23034, (Mouse) 74480, (Rat) 305826
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.