Abnova
Manufacturer Code:H00148398K
Catalog # 89-208-865
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human SAMD11 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human SAMD11 peptide using ARM Technology.
For Research Use Only
Protein Aliases: MGC45873; SAM domain-containing protein 11; sterile alpha motif domain containing 11 splice variant ASV1; sterile alpha motif domain containing 11 splice variant ASV10; sterile alpha motif domain containing 11 splice variant ASV11; sterile alpha motif domain containing 11 splice variant ASV12; sterile alpha motif domain containing 11 splice variant ASV13; sterile alpha motif domain containing 11 splice variant ASV14; sterile alpha motif domain containing 11 splice variant ASV15; sterile alpha motif domain containing 11 splice variant ASV16; sterile alpha motif domain containing 11 splice variant ASV17; sterile alpha motif domain containing 11 splice variant ASV18; sterile alpha motif domain containing 11 splice variant ASV19; sterile alpha motif domain containing 11 splice variant ASV2; sterile alpha motif domain containing 11 splice variant ASV20; sterile alpha motif domain containing 11 splice variant ASV21; sterile alpha motif domain containing 11 splice variant ASV22; sterile alpha motif domain containing 11 splice variant ASV23; sterile alpha motif domain containing 11 splice variant ASV24; sterile alpha motif domain containing 11 splice variant ASV25; sterile alpha motif domain containing 11 splice variant ASV26; sterile alpha motif domain containing 11 splice variant ASV27; sterile alpha motif domain containing 11 splice variant ASV28; sterile alpha motif domain containing 11 splice variant ASV29; sterile alpha motif domain containing 11 splice variant ASV3; sterile alpha motif domain containing 11 splice variant ASV30; sterile alpha motif domain containing 11 splice variant ASV31; sterile alpha motif domain containing 11 splice variant ASV32; sterile alpha motif domain containing 11 splice variant ASV33; sterile alpha motif domain containing 11 splice variant ASV34; sterile alpha motif domain containing 11 splice variant ASV35; sterile alpha motif domain containing 11 splice variant ASV36; sterile alpha motif domain containing 11 splice variant ASV37; sterile alpha motif domain containing 11 splice variant ASV38; sterile alpha motif domain containing 11 splice variant ASV39; sterile alpha motif domain containing 11 splice variant ASV4; sterile alpha motif domain containing 11 splice variant ASV40; sterile alpha motif domain containing 11 splice variant ASV41; sterile alpha motif domain containing 11 splice variant ASV42; sterile alpha motif domain containing 11 splice variant ASV43; sterile alpha motif domain containing 11 splice variant ASV44; sterile alpha motif domain containing 11 splice variant ASV45; sterile alpha motif domain containing 11 splice variant ASV5; sterile alpha motif domain containing 11 splice variant ASV6; sterile alpha motif domain containing 11 splice variant ASV7; sterile alpha motif domain containing 11 splice variant ASV8; sterile alpha motif domain containing 11 splice variant ASV9; Sterile alpha motif domain-containing protein 11
Gene Aliases: MRS; SAMD11
UniProt ID: (Human) Q96NU1
Entrez Gene ID: (Human) 148398
Molecular Function: DNA binding protein chromatin/chromatin-binding protein nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.