Invitrogen
Manufacturer Code:PA596507
Catalog # PIPA596507
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 300 to the C-terminus of human KHDRBS1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GAP-associated tyrosine phosphoprotein p62; GAP-associated tyrosine phosphoprotein p62 (Sam68); KH domain containing, RNA binding, signal transduction associated 1; KH domain-containing, RNA-binding, signal transduction-associated protein 1; KHDRBS1; khdrbs1 {ECO:0000312|RGD:621459}; p21 Ras GTPase-activating protein-associated p62; p68; Sam68; src associated in mitosis, 68 kDa; Src-associated in mitosis 68 kDa protein
Gene Aliases: KHDRBS1; P62; p68; SAM68
UniProt ID: (Human) Q07666, (Mouse) Q60749, (Rat) Q91V33
Entrez Gene ID: (Human) 10657, (Mouse) 20218, (Rat) 117268
Molecular Function: RNA binding protein mRNA processing factor mRNA splicing factor nucleic acid binding transcription cofactor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.