Invitrogen
Manufacturer Code:PA530400
Catalog # PIPA530400
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 104 and 412 of Human SAM68 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GAP-associated tyrosine phosphoprotein p62; GAP-associated tyrosine phosphoprotein p62 (Sam68); KH domain containing, RNA binding, signal transduction associated 1; KH domain-containing, RNA-binding, signal transduction-associated protein 1; KHDRBS1; khdrbs1 {ECO:0000312|RGD:621459}; p21 Ras GTPase-activating protein-associated p62; p68; Sam68; Sam68 p62 p68 GAP-associated tyrosine phosphoprotein p62 (Sam68) KH domain-containing RNA-binding signal transduction-associated protein 1 p21 Ras GTPase-activating protein-associated p62 src-associated in mitosis 68 kDa protein; src associated in mitosis, 68 kDa; Src-associated in mitosis 68 kDa protein
Gene Aliases: KHDRBS1; P62; p68; SAM68
UniProt ID: (Human) Q07666, (Rat) Q91V33
Entrez Gene ID: (Human) 10657, (Rat) 117268
Molecular Function:
RNA binding protein
mRNA processing factor
mRNA splicing factor
nucleic acid binding
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.