Invitrogen
Manufacturer Code:PA5102929
Catalog # PIPA5102929
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human KHDRBS1 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ34027 GAP-associated tyrosine phosphoprotein p62 GAP-associated tyrosine phosphoprotein p62 (Sam68) KH domain containing RNA binding signal transduction associated 1 KH domain-containing RNA-binding signal transduction-associated protein 1 KH RNA binding domain containing signal transduction associated 1 Khdrbs1 khdrbs1 {ECO:0000312 p21 Ras GTPase-activating protein-associated p62 P62 p68 RGD:621459} Sam68 src associated in mitosis 68 kDa Src-associated in mitosis 68 kDa protein; GAP-associated tyrosine phosphoprotein p62; GAP-associated tyrosine phosphoprotein p62 (Sam68); KH domain containing, RNA binding, signal transduction associated 1; KH domain-containing, RNA-binding, signal transduction-associated protein 1; KHDRBS1; p21 Ras GTPase-activating protein-associated p62; p68; Sam68; src associated in mitosis, 68 kDa; Src-associated in mitosis 68 kDa protein
Gene Aliases: KHDRBS1; p62; p68; SAM68
UniProt ID: (Human) Q07666, (Mouse) Q60749
Entrez Gene ID: (Human) 10657, (Mouse) 20218
Molecular Function:
RNA binding protein
mRNA processing factor
mRNA splicing factor
nucleic acid binding
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.