Novus Biologicals
Manufacturer Code:NBP220254
Catalog # NBP220254
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human SAM68. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ34027 GAP-associated tyrosine phosphoprotein p62 KH domain containing RNA binding signal transduction associated 1 KH domain-containing RNA-binding signal transduction-associated protein 1 p21 Ras GTPase-activating protein-associated p62 p62 p68 SAM68 Sam68GAP-associated tyrosine phosphoprotein p62 (Sam68) Src-associated in mitosis 68 kDa protein; GAP-associated tyrosine phosphoprotein p62; GAP-associated tyrosine phosphoprotein p62 (Sam68); KH domain containing, RNA binding, signal transduction associated 1; KH domain-containing, RNA-binding, signal transduction-associated protein 1; KHDRBS1; p21 Ras GTPase-activating protein-associated p62; p68; Sam68; Src-associated in mitosis 68 kDa protein
Gene Aliases: KHDRBS1; p62; p68; SAM68
UniProt ID: (Human) Q07666
Entrez Gene ID: (Human) 10657
Molecular Function:
RNA binding protein
mRNA processing factor
mRNA splicing factor
nucleic acid binding
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.