Invitrogen
Manufacturer Code:PA5115864
Catalog # PIPA5115864
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human SALL1 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: epididymis secretory protein Li 89; epididymis secretory protein Li 89 HEL-S-89 HSAL1 Msal-3 SAL1 Sal-1 sal-3 SALL1 sal-like 1 (Drosophila) sal-like protein 1 spalt like transcription factor 1 spalt-like transcription factor 1 TBS Zinc finger protein 794 zinc finger protein SALL1 Zinc finger protein Spalt-1 zinc finger protein Spalt-3 ZNF794; HSal1; sal-3; sal-like 1; Sal-like protein 1; Spalt-like transcription factor 1; Zinc finger protein 794; Zinc finger protein SALL1; Zinc finger protein Spalt-1; zinc finger protein Spalt-3
Gene Aliases: HEL-S-89; HSAL1; Msal-3; Sal-1; SAL1; SALL1; TBS; ZNF794
UniProt ID: (Human) Q9NSC2
Entrez Gene ID: (Human) 6299, (Rat) 307740, (Mouse) 58198
Molecular Function:
nucleic acid binding
transcription factor
zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.