Invitrogen
Manufacturer Code:PA567080
Catalog # PIPA567080
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human SACM1L |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Phosphatidylinositol-3-phosphatase SAC1; Phosphatidylinositol-4-phosphate phosphatase; SAC1 phosphatidylinositide phosphatase SAC1 suppressor of actin 1 suppressor of actin mutations 1-like protein; suppressor of actin 1; Suppressor of actin mutations 1-like protein
Gene Aliases: KIAA0851; SAC1; SACM1L
UniProt ID: (Human) Q9NTJ5
Entrez Gene ID: (Human) 22908
Molecular Function:
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.