Invitrogen
Manufacturer Code:PA542961
Catalog # PIPA542961
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human SACM1L |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: phosphatidylinositide phosphatase SAC1; Phosphatidylinositol-3-phosphatase SAC1; Phosphatidylinositol-4-phosphate phosphatase; polyphosphatidylinositol phosphatase; SAC1 suppressor of actin mutations 1 like; suppressor of actin 1; Suppressor of actin mutations 1-like protein
Gene Aliases: KIAA0851; mKIAA0851; SAC1; Sac1p; SACM1L
UniProt ID: (Bovine) A6QL88, (Human) Q9NTJ5, (Mouse) Q9EP69, (Rat) Q9ES21
Entrez Gene ID: (Bovine) 530577, (Horse) 100065601, (Dog) 484795, (Rabbit) 100347235, (Human) 22908, (Mouse) 83493, (Rat) 116482, (Guinea pig) 101787561
Molecular Function: hydrolase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.