Invitrogen
Manufacturer Code:PA5106374
Catalog # PIPA5106374
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human SACM1L corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Kiaa0851 mKIAA0851 Phosphatidylinositide phosphatase SAC1 polyphosphatidylinositol phosphatase Sac1 SAC1 (suppressor of actin mutations 1 homolog)-like (S. cerevisiae) SAC1 suppressor of actin mutations 1-like (yeast) Sac1p SACM1L suppressor of actin 1 suppressor of actin mutations 1-like protein; phosphatidylinositide phosphatase SAC1; Phosphatidylinositol-3-phosphatase SAC1; Phosphatidylinositol-4-phosphate phosphatase; polyphosphatidylinositol phosphatase; suppressor of actin 1; Suppressor of actin mutations 1-like protein
Gene Aliases: KIAA0851; mKIAA0851; SAC1; Sac1p; SACM1L
UniProt ID: (Human) Q9NTJ5, (Mouse) Q9EP69, (Rat) Q9ES21
Entrez Gene ID: (Human) 22908, (Mouse) 83493, (Rat) 116482
Molecular Function: hydrolase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.