Abnova
Manufacturer Code:RAB00624
Catalog # 89-339-603
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | SAA/2868R |
Immunogen | Recombinant protein corresponding to full-length human SAA |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20 to -80°C |
Rabbit Monoclonal Antibody.
Protein Aliases: AA2; Amyloid A2 protein; Amyloid fibril protein AA; Amyloid protein A; MGC111216 PIG4 SAA TP53I4; SAA; SAA2; serum amyloid A protein; Serum amyloid A-1 protein; Serum amyloid A-2 protein; Serum amyloid protein A(2-102); Serum amyloid protein A(2-103); Serum amyloid protein A(2-104); Serum amyloid protein A(3-104); Serum amyloid protein A(4-101); tumor protein p53 inducible protein 4
Gene Aliases: PIG4; SAA; SAA1; SAA2; TP53I4
UniProt ID: (Human) P0DJI8, (Human) P0DJI9
Entrez Gene ID: (Human) 6288, (Human) 6289
Molecular Function:
apolipoprotein
defense/immunity protein
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.