Abnova
Manufacturer Code:H00006288K
Catalog # 89-210-173
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human SAA1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human SAA1 peptide using ARM Technology.
For Research Use Only
Protein Aliases: Amyloid fibril protein AA; Amyloid protein A; MGC111216 PIG4 SAA TP53I4; SAA; serum amyloid A protein; Serum amyloid A-1 protein; Serum amyloid protein A(2-102); Serum amyloid protein A(2-103); Serum amyloid protein A(2-104); Serum amyloid protein A(3-104); Serum amyloid protein A(4-101); tumor protein p53 inducible protein 4
Gene Aliases: PIG4; SAA; SAA1; SAA2; TP53I4
UniProt ID: (Human) P0DJI8
Entrez Gene ID: (Human) 6288
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.