Proteintech
Manufacturer Code:138741AP150UL
Catalog # 50-172-9058
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | S1PR5/EDG8 Fusion Protein Ag4928 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Edg 8 EDG8 S1P receptor 5 S1P receptor Edg 8 S1P5 S1PR5 SPPR 1 SPPR 2; Endothelial differentiation G-protein-coupled receptor 8; endothelial differentiation, sphingolipid G-protein-coupled receptor, 8; Lysophospholipid receptor B4; Nerve growth factor-regulated G-protein-coupled receptor 1; NRG-1; S1P receptor 5; S1P receptor Edg-8; S1P5; Sphingosine 1-phosphate receptor 5; Sphingosine 1-phosphate receptor Edg-8; sphingosine 1-phosphate receptor EDG8; sphingosine-1-phosphate receptor LPB4
Gene Aliases: Edg-8; EDG8; Lpb4; Nrg1; S1P5; S1PR5; SPPR-1; SPPR-2
UniProt ID: (Human) Q9H228, (Rat) Q9JKM5, (Mouse) Q91X56
Entrez Gene ID: (Human) 53637, (Rat) 60399, (Mouse) 94226
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.