Abnova
Manufacturer Code:PAB26238
Catalog # 89-127-879
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 19 amino acid at N-terminus of human S1PR3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of S1PR3.
For Research Use Only
Protein Aliases: EDG-3; EDG3; Endothelial differentiation G-protein coupled receptor 3; endothelial differentiation, sphingolipid G-protein-coupled receptor, 3; FLJ37523; FLJ93220; G protein-coupled receptor, endothelial differentiation gene-3; LPB3; MGC71696; S1P receptor 3; S1P receptor Edg-3; S1P receptor EDG3; S1P3; Sphingosine 1-phosphate receptor 3; Sphingosine 1-phosphate receptor Edg-3
Gene Aliases: EDG-3; EDG3; LPB3; S1P3; S1PR3
UniProt ID: (Human) Q99500
Entrez Gene ID: (Human) 1903
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.