R&D Systems
Manufacturer Code:FAB7524S100UG
Catalog # FAB7524S100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 776808 |
Immunogen | Human S1P3/EDG-3 transfected NS0 cells Accession # Q99500 |
Conjugate | Alexa Fluor 750 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Mouse MonoclonalAntibody
For Research Use Only
Protein Aliases: EDG3 EDG-3 EDG3FLJ37523 Endothelial differentiation G-protein coupled receptor 3 endothelial differentiation sphingolipid G-protein-coupled receptor 3 FLJ93220 G protein-coupled receptor endothelial differentiation gene-3 LPB3 MGC71696 S1P receptor 3 S1P receptor Edg-3 S1P receptor EDG3 S1P3 S1PR3 sphingosine 1-phosphate receptor 3 Sphingosine 1-phosphate receptor Edg-3 sphingosine-1-phosphate receptor 3; Endothelial differentiation G-protein coupled receptor 3; endothelial differentiation, sphingolipid G-protein-coupled receptor, 3; G protein-coupled receptor, endothelial differentiation gene-3; S1P receptor 3; S1P receptor Edg-3; S1P receptor EDG3; Sphingosine 1-phosphate receptor 3; Sphingosine 1-phosphate receptor Edg-3
Gene Aliases: EDG-3; EDG3; LPB3; S1P3; S1PR3
UniProt ID: (Human) Q99500
Entrez Gene ID: (Human) 1903
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.