R&D Systems
Manufacturer Code:FAB2016G100UG
Catalog # FAB2016G100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 218713 |
Immunogen | NS0 mouse myeloma cell line transfected with human S1P 1EDG-1 Met1-Ser382 Accession # P21453 |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Mouse MonoclonalAntibody
For Research Use Only
Protein Aliases: CD363; CD363 CD363 antigen CHEDG1 D1S3362 EDG1 EDG-1 EDG1edg-1 Endothelial differentiation G-protein coupled receptor 1 endothelial differentiation sphingolipid G-protein-coupled receptor 1 FLJ58121 S1P receptor 1 S1P receptor Edg-1 s1p1 S1P1ECGF1 S1PR1 sphingosine 1-phosphate receptor 1 sphingosine 1-phosphate receptor EDG1 Sphingosine 1-phosphate receptor Edg-1 sphingosine-1-phosphate receptor 1; Endothelial differentiation G-protein coupled receptor 1; endothelial differentiation, sphingolipid G-protein-coupled receptor, 1; S1P receptor 1; S1P receptor Edg-1; Sphingosine 1-phosphate receptor 1; Sphingosine 1-phosphate receptor Edg-1; sphingosine 1-phosphate receptor EDG1
Gene Aliases: CD363; CHEDG1; D1S3362; ECGF1; EDG-1; EDG1; S1P1; S1PR1
UniProt ID: (Human) P21453
Entrez Gene ID: (Human) 1901
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.