Novus Biologicals
Manufacturer Code:NBP224673SS
Catalog # NBP224673SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A portion of amino acids 350 to 400 of human S1PR1 was used as immunogen for this antibody. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD363; CD363 CD363 antigen CHEDG1 D1S3362 EDG-1 EDG1edg-1 Endothelial differentiation G-protein coupled receptor 1 endothelial differentiation sphingolipid G-protein-coupled receptor 1 FLJ58121 S1P receptor 1 S1P receptor Edg-1 s1p1 S1P1ECGF1 sphingosine 1-phosphate receptor 1 sphingosine 1-phosphate receptor EDG1 Sphingosine 1-phosphate receptor Edg-1 sphingosine-1-phosphate receptor 1; Endothelial differentiation G-protein coupled receptor 1; endothelial differentiation, sphingolipid G-protein-coupled receptor, 1; S1P receptor 1; S1P receptor Edg-1; Sphingosine 1-phosphate receptor 1; Sphingosine 1-phosphate receptor Edg-1; sphingosine 1-phosphate receptor EDG1
Gene Aliases: CD363; CHEDG1; D1S3362; ECGF1; EDG-1; EDG1; S1P1; S1PR1
UniProt ID: (Human) P21453
Entrez Gene ID: (Human) 1901
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.