Invitrogen
Manufacturer Code:MA535431
Catalog # PIMA535431
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC0881 |
Immunogen | A synthesized peptide derived from human EDG1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: AI849002 CD antigen CD363 CD363 CD363 antigen CHEDG1 D1S3362 ECGF1 Edg1 EDG-1 Endothelial differentiation G-protein coupled receptor 1 etID43307.22 Lpb1 Lysophospholipid receptor B1 S1p S1P receptor 1 S1P receptor Edg-1 s1p1 S1pr1 Sphingosine 1-phosphate receptor 1 Sphingosine 1-phosphate receptor Edg-1 wu:fb98f05; CD363; EDG1 (Edg1); Endothelial differentiation G-protein coupled receptor 1; endothelial differentiation sphingolipid G-protein-coupled receptor 1; endothelial differentiation, sphingolipid G-protein-coupled receptor, 1; Lysophospholipid receptor B1; S1P receptor 1; S1P receptor Edg-1; S1P1; Sphingosine 1-phosphate receptor 1; Sphingosine 1-phosphate receptor Edg-1; sphingosine 1-phosphate receptor EDG1
Gene Aliases: AI849002; CD363; CHEDG1; D1S3362; ECGF1; EDG-1; EDG1; Lpb1; S1p; S1P1; S1PR1
UniProt ID: (Human) P21453, (Rat) P48303, (Mouse) O08530
Entrez Gene ID: (Human) 1901, (Rat) 29733, (Mouse) 13609
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.