Invitrogen
Manufacturer Code:PA585068
Catalog # PIPA585068
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human EDG1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD363; CHEDG1 antibody D1S3362 antibody ECGF1 antibody EDG-1 antibody EDG1 antibody FLJ58121 antibody S1P1 antibody S1PR1 antibody sphingosine 1-phosphate receptor Edg-1 antibody endothelial differentiation sphingolipid G-protein-coupled receptor 1 antibody S1P receptor 1 antibody endothelial differentiation G-protein coupled receptor 1 antibody S1P receptor Edg-1 antibody sphingosine 1-phosphate receptor EDG1 antibody sphingosine 1-phosphate receptor 1 antibody sphingosine-1-phosphate receptor 1 antibody; Endothelial differentiation G-protein coupled receptor 1; endothelial differentiation sphingolipid G-protein-coupled receptor 1; endothelial differentiation, sphingolipid G-protein-coupled receptor, 1; Lysophospholipid receptor B1; S1P receptor 1; S1P receptor Edg-1; Sphingosine 1-phosphate receptor 1; Sphingosine 1-phosphate receptor Edg-1; sphingosine 1-phosphate receptor EDG1
Gene Aliases: AI849002; CD363; CHEDG1; D1S3362; ECGF1; EDG-1; EDG1; Lpb1; S1p; S1P1; S1PR1
UniProt ID: (Human) P21453, (Mouse) O08530
Entrez Gene ID: (Human) 1901, (Mouse) 13609
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.