Invitrogen
Manufacturer Code:PA572648
Catalog # PIPA572648
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Zebrafish |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 311-335 amino acids from the C-terminal region of zebrafish S1P1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Edg1; rhodopsin family G protein-coupled receptor; endothelial differentiation, sphingolipid G-protein-coupled receptor, 1; S1P receptor 1; S1P receptor Edg-1; Sphingosine 1-phosphate receptor 1; Sphingosine 1-phosphate receptor 1 S1P receptor 1 S1P1 Sphingosine 1-phosphate receptor Edg-1 S1P receptor Edg-1 s1pr1 edg1; Sphingosine 1-phosphate receptor Edg-1
Gene Aliases: edg1; etID43307.22; s1p1; s1pr1; wu:fb98f05
UniProt ID: (Zebrafish) Q9DDK4
Entrez Gene ID: (Zebrafish) 64617
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.