Invitrogen
Manufacturer Code:PA534180
Catalog # PIPA534180
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 19 amino acid peptide from C-terminus of human S1PR1 / EDG1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD363; Endothelial differentiation G-protein coupled receptor 1; endothelial differentiation, sphingolipid G-protein-coupled receptor, 1; S1P receptor 1; S1P receptor Edg-1; S1PR1 CD363 CD363 antigen CHEDG1 ECGF1 EDG-1 D1S3362 EDG1 S1P1 S1P receptor Edg-1 S1P receptor 1; Sphingosine 1-phosphate receptor 1; Sphingosine 1-phosphate receptor Edg-1; sphingosine 1-phosphate receptor EDG1
Gene Aliases: CD363; CHEDG1; D1S3362; ECGF1; EDG-1; EDG1; S1P1; S1PR1
UniProt ID: (Human) P21453
Entrez Gene ID: (Human) 1901
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.