Invitrogen
Manufacturer Code:PA524124
Catalog # PIPA524124
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human S100Z |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Gm625 S100-zeta S100 calcium binding protein zeta S100 calcium-binding protein Z protein S100-Z; Protein S100-Z; S100 calcium binding protein, zeta; S100 calcium-binding protein Z
Gene Aliases: Gm625; S100-zeta; S100Z
UniProt ID: (Human) Q8WXG8
Entrez Gene ID: (Human) 170591, (Mouse) 268686
Molecular Function: calcium-binding protein calmodulin intracellular calcium-sensing protein signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.