Invitrogen
Manufacturer Code:PA5101793
Catalog # PIPA5101793
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human S100Z corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Gm625 protein S100-Z S100 calcium binding protein Z S100 calcium binding protein zeta S100 calcium-binding protein Z S100Z S100-zeta; Protein S100-Z; S100 calcium binding protein, zeta; S100 calcium-binding protein Z
Gene Aliases: Gm625; S100-zeta; S100Z
UniProt ID: (Human) Q8WXG8
Entrez Gene ID: (Human) 170591
Molecular Function: calcium-binding protein calmodulin intracellular calcium-sensing protein signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.