R&D Systems
Manufacturer Code:MAB18202100
Catalog # MAB18202100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 721713 |
Immunogen | <i>E. coli</i>-derived recombinant human S100B Met1-Glu92 Accession &num P04271 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70°C as supplied. 1 month 2 to 8°C under sterile conditions after reconstitution. 6 months -20 to -70°C under sterile conditions a |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: beta (neural) NEF S100 S100 beta S100 calcium binding protein B S100 calcium-binding protein B S100 calcium-binding protein beta (neural) S-100 calcium-binding protein beta chain 10protein S100-B S-100 protein beta chain S-100 protein subunit beta S100beta; Protein S100-B; S-100 calcium-binding protein, beta chain; S-100 protein beta chain; S-100 protein subunit beta; S100 calcium-binding protein B; S100 calcium-binding protein, beta (neural)
Gene Aliases: NEF; S100; S100-B; S100B; S100beta
UniProt ID: (Human) P04271
Entrez Gene ID: (Human) 6285
Molecular Function:
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.