Novus Biologicals
Manufacturer Code:NBP245224
Catalog # NBP245224
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 15F4NB |
Immunogen | Partial recombinant human S100B protein (amino acids 2-92) [UniProt P04271] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: beta (neural) NEF S100 S100 beta S100 calcium binding protein B S100 calcium-binding protein B S100 calcium-binding protein beta (neural) S-100 calcium-binding protein beta chain 10protein S100-B S-100 protein beta chain S-100 protein subunit beta S100beta; Protein S100-B; S-100 calcium-binding protein, beta chain; S-100 protein beta chain; S-100 protein subunit beta; S100 calcium-binding protein B; S100 calcium-binding protein, beta (neural)
Gene Aliases: NEF; S100; S100-B; S100B; S100beta
UniProt ID: (Human) P04271
Entrez Gene ID: (Human) 6285
Molecular Function:
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.