Proteintech
Manufacturer Code:142261AP150UL
Catalog # 50-556-207
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | S100A9 Fusion Protein Ag5465 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 60B8AG CAGB Calgranulin B Calprotectin L1H subunit CFAG CGLB L1AG LIAG MAC387 MIF MRP 14 MRP14 NIF P14 Protein S100 A9 S100A9; calgranulin B; Calgranulin-B; Calprotectin L1H subunit; intracellular calcium-binding protein (MRP14); Leukocyte L1 complex heavy chain; Migration inhibitory factor-related protein 14; MRP-14; Myeloid-related protein 14; p14; Protein S100-A9; S100 calcium binding protein A9 (calgranulin B); S100 calcium-binding protein A9; S100 calcium-binding protein A9 (calgranulin B)
Gene Aliases: 60B8AG; AW546964; BEE22; CAGB; CFAG; CGLB; GAGB; L1AG; LIAG; MAC387; MIF; MRP14; NIF; P14; S100A9
UniProt ID: (Human) P06702, (Mouse) P31725, (Rat) P50116
Entrez Gene ID: (Human) 6280, (Mouse) 20202, (Rat) 94195
Molecular Function: calcium-binding protein calmodulin intracellular calcium-sensing protein signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.