R&D Systems
Manufacturer Code:MAB4570
Catalog # MAB4570
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 749916 |
Immunogen | <EM>E. coli</EM>-derived recombinant human S100A8 Met1-Glu93 Accession # P05109 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 60B8AG CAGACP-10 calgranulin A Calgranulin-A Calprotectin L1L subunit CFAG CFAGL1Ag CGLA Cystic fibrosis antigen Leukocyte L1 complex light chain MA387 MIF Migration inhibitory factor-related protein 8 MRP8 MRP-8 MRP8S100 calcium binding protein A8 (calgranulin A) NIF P8 protein S100-A8 S100 calcium binding protein A8 S100 calcium-binding protein A8 S100 calcium-binding protein A8 (calgranulin A) Urinary stone protein band A; calgranulin A; Calgranulin-A; Calprotectin L1L subunit; CFAG; Cystic fibrosis antigen; Leukocyte L1 complex light chain; Migration inhibitory factor-related protein 8; MRP-8; Protein S100-A8; S100 calcium-binding protein A8; S100 calcium-binding protein A8 (calgranulin A); Urinary stone protein band A
Gene Aliases: 60B8AG; CAGA; CFAG; CGLA; CP-10; L1Ag; MA387; MIF; MRP8; NIF; P8; S100A8
UniProt ID: (Human) P05109
Entrez Gene ID: (Human) 6279
Molecular Function: calcium-binding protein calmodulin intracellular calcium-sensing protein signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.