Invitrogen
Manufacturer Code:PA579948
Catalog # PIPA579948
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E.coli-derived mouse S100A8 recombit protein (Position: P2-E89). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Calgranulin-A; Chemotactic cytokine CP-10; chemotactic S100 protein; Leukocyte L1 complex light chain; Migration inhibitory factor-related protein 8; MRP-8; p8; Pro-inflammatory S100 cytokine; Protein S100-A8; Protein S100-A8 Calgranulin-A Chemotactic cytokine CP-10 Leukocyte L1 complex light chain Migration inhibitory factor-related protein 8 MRP-8 p8 Pro-inflammatory S100 cytokine S100 calcium-binding protein A8 S100a8 Caga Mrp8; S100 calcium binding protein A8 (calgranulin A); S100 calcium-binding protein A8; S100 calcium-binding protein A8 (calgranulin A)
Gene Aliases: 60B8Ag; AI323541; B8Ag; Caga; CFAg; CP-10; MRP8; p8; S100a8
UniProt ID: (Rat) P50115, (Mouse) P27005
Entrez Gene ID: (Rat) 116547, (Mouse) 20201
Molecular Function: calcium-binding protein calmodulin intracellular calcium-sensing protein signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.