Invitrogen
Manufacturer Code:PA547439
Catalog # PIPA547439
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived recombit mouse S100A8 Pro2-Glu89 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: CAGA CFAG MRP8 Protein S100-A8 S100A8 Calprotectin L1L subunit Cystic fibrosis antigen CFAG Leukocyte L1 complex light chain Migration inhibitory factor-related protein 8 P8; Calgranulin-A; Chemotactic cytokine CP-10; chemotactic S100 protein; Leukocyte L1 complex light chain; Migration inhibitory factor-related protein 8; MRP-8; p8; Pro-inflammatory S100 cytokine; Protein S100-A8; S100 calcium binding protein A8 (calgranulin A); S100 calcium-binding protein A8; S100 calcium-binding protein A8 (calgranulin A)
Gene Aliases: 60B8Ag; AI323541; B8Ag; Caga; CFAg; CP-10; MRP8; p8; S100a8
UniProt ID: (Rat) P50115, (Mouse) P27005
Entrez Gene ID: (Rat) 116547, (Mouse) 20201
Molecular Function: calcium-binding protein calmodulin intracellular calcium-sensing protein signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.