Invitrogen
Manufacturer Code:PA570701
Catalog # PIPA570701
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human S100A7A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Protein S100-A7A; S100 calcium binding protein A15; S100 calcium binding protein A7-like 1; S100 calcium-binding protein A15; S100 calcium-binding protein A7-like 1; S100 calcium-binding protein A7A; S100A15 S100A7L1
Gene Aliases: NICE-2; S100A15; S100A7A; S100A7f; S100A7L1
UniProt ID: (Human) Q86SG5
Entrez Gene ID: (Goat) 102173556, (Human) 338324
Molecular Function: calcium-binding protein calmodulin intracellular calcium-sensing protein signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.