Novus Biologicals
Manufacturer Code:NBP28981450UL
Catalog # NB021098
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 129 |
Immunogen | Recombinant Human S100A7/Psoriasin protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: HID 5 HID5 HID-5 protein S100-A7 PSOR1 Psoriasin psoriasin 1 S100 calcium binding protein A7 S100 calcium binding protein A7 (psoriasin 1) S100 calcium-binding protein A7 S100 calcium-binding protein A7 (psoriasin 1) S100A7c; Protein S100-A7; Psoriasin; psoriasin 1; S100 calcium-binding protein A7; S100 calcium-binding protein A7 (psoriasin 1)
Gene Aliases: PSOR1; S100A7; S100A7C
UniProt ID: (Human) P31151
Entrez Gene ID: (Human) 6278
Molecular Function: calcium-binding protein calmodulin intracellular calcium-sensing protein signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.