Invitrogen
Manufacturer Code:PA579947
Catalog # PIPA579947
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived human Psoriasin recombit protein (Position: S2-Q101). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Protein S100-A15A; Protein S100-A7; Protein S100-A7 Psoriasin S100 calcium-binding protein A7 S100A7 PSOR1 S100A7C; Protein S100-A7A; Psoriasin; psoriasin 1; S100 calcium binding protein A15; S100 calcium binding protein A7-like 1; S100 calcium-binding protein A15; S100 calcium-binding protein A15A; S100 calcium-binding protein A7; S100 calcium-binding protein A7 (psoriasin 1); S100 calcium-binding protein A7A
Gene Aliases: AY465109; Gm1020; PSOR1; S100a15; S100a15a; S100a17l1; S100A7; S100a7a; S100A7C; S100A7f; S100A7L1
UniProt ID: (Human) P31151, (Mouse) Q6S5I3
Entrez Gene ID: (Human) 6278, (Mouse) 381493
Molecular Function: calcium-binding protein calmodulin intracellular calcium-sensing protein signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.