Invitrogen
Manufacturer Code:PA5116994
Catalog # PIPA5116994
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-90 of human S100A6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2A9 5B10 CABP Cacy calcium binding protein A6 (calcyclin) calcyclin Growth factor-inducible protein 2A9 lung 10 kDa protein MLN 4 PRA Prolactin receptor-associated protein protein S100-A6 S100 calcium binding protein A6 S100 calcium binding protein A6 (calcyclin) S100 calcium-binding protein A6 S100 calcium-binding protein A6 (calcyclin) S100a6; 5B10; calcium binding protein A6 (calcyclin); Calcyclin; Growth factor-inducible protein 2A9; MLN 4; PRA; Prolactin receptor-associated protein; Protein S100-A6; S100 calcium-binding protein A6; S100 calcium-binding protein A6 (calcyclin)
Gene Aliases: 2A9; 5B10; CABP; CACY; CALCYCLIN; PRA; S100A6
UniProt ID: (Human) P06703, (Mouse) P14069, (Rat) P05964
Entrez Gene ID: (Human) 6277, (Mouse) 20200, (Rat) 85247
Molecular Function:
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.