Novus Biologicals
Manufacturer Code:NBP290583FR
Catalog # NB075271
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 1 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Mouse S100A3 (NP_035440.1 Met1-Gln101). |
Conjugate | DyLight 680 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: Protein S-100E; Protein S-100E S100 calcium binding protein A3 S100 calcium-binding protein A3S100Eprotein S100-A3; Protein S100-A3; S100 calcium-binding protein A3
Gene Aliases: S100A3; S100E
UniProt ID: (Human) P33764
Entrez Gene ID: (Human) 6274
Molecular Function: calcium-binding protein calmodulin growth factor intracellular calcium-sensing protein signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.