Invitrogen
Manufacturer Code:PA542097
Catalog # PIPA542097
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human S100A3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Protein S-100E; Protein S100-A3; S100 Calcium binding protein A3 S100E; S100 calcium-binding protein A3
Gene Aliases: S100A3; S100E
UniProt ID: (Human) P33764, (Rat) P62819, (Mouse) P62818
Entrez Gene ID: (Dog) 490460, (Human) 6274, (Rabbit) 100347250, (Horse) 100061923, (Bovine) 786230, (Rat) 114216, (Guinea pig) 100724226, (Mouse) 20197
Molecular Function:
calcium-binding protein
calmodulin
growth factor
intracellular calcium-sensing protein
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.