Invitrogen
Manufacturer Code:PA5103030
Catalog # PIPA5103030
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human S100A2 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CAN19; CAN19 EG628324 predicted gene EG628324 Protein S100-A2 Protein S-100L S100 calcium binding protein A2 S100 calcium-binding protein A2 S100A2 S100L; Protein S-100L; Protein S100-A2; S100 calcium-binding protein A2
Gene Aliases: CAN19; S100A2; S100L
UniProt ID: (Human) P29034
Entrez Gene ID: (Human) 6273
Molecular Function: calcium-binding protein calmodulin growth factor intracellular calcium-sensing protein signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.