Abnova
Manufacturer Code:PAB19570
Catalog # 89-121-097
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to full length human S100A2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against full-length recombinant S100A2.
For Research Use Only
Protein Aliases: CAN19; MGC111539; Protein S-100L; Protein S100-A2; S100 calcium-binding protein A2; S100L
Gene Aliases: CAN19; S100A2; S100L
UniProt ID: (Human) P29034
Entrez Gene ID: (Human) 6273
Molecular Function: calcium-binding protein calmodulin growth factor intracellular calcium-sensing protein signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.