Proteintech
Manufacturer Code:102371AP150UL
Catalog # 50-172-6139
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | S100A11 Fusion Protein Ag0343 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Calgizzarin; Calgizzarin MLN 70 MLN70 Protein S100 A11 Protein S100 C S100A11 S100C; calizzarin; EMAP; endothelial monocyte activating polypeptide 1; Endothelial monocyte-activating polypeptide; epididymis secretory protein Li 43; Metastatic lymph node gene 70 protein; MLN 70; Protein S100-A11; Protein S100-A11, N-terminally processed; Protein S100-C; S100 calcium binding protein A11 (calizzarin); S100 calcium binding protein A14; S100 calcium-binding protein A11; S100 calcium-binding protein A11 (calgizzarin)
Gene Aliases: cal; Emap1; EMAPI; HEL-S-43; MLN70; S100A11; S100a14; S100C
UniProt ID: (Human) P31949, (Rat) Q6B345, (Mouse) P50543
Entrez Gene ID: (Human) 6282, (Rat) 445415, (Mouse) 20195
Molecular Function: calcium-binding protein calmodulin intracellular calcium-sensing protein signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.